Skip to Content
Merck
All Photos(2)

Key Documents

460060

Sigma-Aldrich

Polyisoprene-graft-maleic anhydride

average Mw ~25,000

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€75.00

€75.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€75.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€75.00


Check Cart for Availability

Request a Bulk Order

mol wt

average Mw ~25,000

refractive index

n20/D 1.52 (lit.)

viscosity

10-50 cP, 20 wt. % in toluene(30 °C, Brookfield)(lit.)

density

0.92 g/mL at 25 °C (lit.)

SMILES string

O1C(=O)C(CC1=O)\C=C\C(=C)C.CC(=C)C=C

InChI key

WRJJKMSOBIKPNE-BJILWQEISA-N

Application

Co-curable plasticizer. Adhesive additive.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the Maleic anhydride content in Polyisoprene-graft-maleic anhydride ??

    1 answer
    1. This product is not tested for maleic anhydride content on a lot by lot basis. The estimated amount is approximately 1.5 wt.%.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service