Skip to Content
Merck
All Photos(1)

Key Documents

455733

Sigma-Aldrich

(2-Methoxyphenoxy)acetic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4OCH2CO2H
CAS Number:
Molecular Weight:
182.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

123-126 °C (lit.)

SMILES string

COc1ccccc1OCC(O)=O

InChI

1S/C9H10O4/c1-12-7-4-2-3-5-8(7)13-6-9(10)11/h2-5H,6H2,1H3,(H,10,11)

InChI key

IHONYPFTXGQWAX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Chen et al.
Applied and environmental microbiology, 50(6), 1451-1456 (1985-12-01)
Veratrylglycerol-beta-guaiacyl ether (0.2 g/liter), a lignin model compound, was found to be degraded by mixed rumen bacteria in a yeast extract medium under strictly anaerobic conditions to the extent of 19% within 24 h. Guaiacoxyacetic acid, 2-(o-methoxyphenoxy)ethanol, vanillic acid, and
H Sasaki et al.
Xenobiotica; the fate of foreign compounds in biological systems, 14(8), 621-631 (1984-08-01)
The disposition and metabolism of amosulalol hydrochloride, a combined alpha- and beta-adrenoceptor blocking agent, were studied in rats, dogs and monkeys. After oral administration of [14C]amosulalol hydrochloride, the plasma concentration of radioactivity reached a maximum at 0.5 to 1 h

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service