Skip to Content
Merck
All Photos(1)

Key Documents

712140

Sigma-Aldrich

Methyl 2,3,4-tri-O-benzyl-α-D-glucopyranoside

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€377.00

€377.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€377.00

About This Item

Empirical Formula (Hill Notation):
C28H32O6
CAS Number:
Molecular Weight:
464.55
Beilstein:
1442065
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

€377.00


Check Cart for Availability

Request a Bulk Order

Assay

≥90% (HPLC)

form

lumps

optical activity

[α]/D 23.0±2.0°, c = 1 in chloroform

storage temp.

2-8°C

SMILES string

CO[C@H]1O[C@H](CO)[C@@H](OCc2ccccc2)[C@H](OCc3ccccc3)[C@H]1OCc4ccccc4

InChI

1S/C28H32O6/c1-30-28-27(33-20-23-15-9-4-10-16-23)26(32-19-22-13-7-3-8-14-22)25(24(17-29)34-28)31-18-21-11-5-2-6-12-21/h2-16,24-29H,17-20H2,1H3/t24-,25-,26+,27-,28+/m1/s1

InChI key

MOKYEUQDXDKNDX-DFLSAPQXSA-N

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service