Skip to Content
Merck

Skip To

8.55140

Fmoc-PAL-Linker

Novabiochem®

Synonym(s):

Fmoc-PAL-Linker

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
€ 85,30
5 g
Check Cart for Availability
€ 366,00

About This Item

Empirical Formula (Hill Notation):
C29H31NO7
CAS Number:
Molecular Weight:
505.56
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

€ 85,30


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

solid

reaction suitability

reagent type: linker

manufacturer/tradename

Novabiochem®

mp

178-180 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

N(Cc4c(cc(cc4OC)OCCCCC(=O)O)OC)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C29H31NO7/c1-34-26-15-19(36-14-8-7-13-28(31)32)16-27(35-2)24(26)17-30-29(33)37-18-25-22-11-5-3-9-20(22)21-10-4-6-12-23(21)25/h3-6,9-12,15-16,25H,7-8,13-14,17-18H2,1-2H3,(H,30,33)(H,31,32)

InChI key

ATYUCXIJDKHOPX-UHFFFAOYSA-N

General description

Barany′s aminomethyl-dimethoxyphenoxyvaleric acid linker . The amino group of this linker can be easily acylated under standard coupling conditions. Following peptide assembly, treatment with 95% TFA containing scavengers releases the desired peptide amide. Studies have shown the acid sensitivity of this linker to be around twice that of the Rink amide linker. There is some evidence to suggest that PAL resins give greater yields in microwave assisted synthesis.

Associated Protocols and Technical Articles


Protocols for Loading of Peptide Synthesis Resins

Analysis Note

Color (visual): white to beige to grey
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(0811)): ≥ 98.0 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.520418.510948.52338
form

solid

form

powder

form

powder

form

powder

functional group

Fmoc

functional group

amine

functional group

Fmoc

functional group

Fmoc

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

storage temp.

2-8°C

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

15-25°C

mp

178-180 °C

mp

170-180 °C

mp

-

mp

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. In the General Description of the product Fmoc-PAL-Linker Catalog 8.55140 there are numbers for two references ([1] and [2]) but I can not either follow a link or see the references. Would it be possible to provide me the references? Thanks

    1 answer
    1. The references for the product Fmoc-PAL-Linker are as follows:

      (1) Gisin BF. 1973. The Preparation of Merrifield-Resins Through Total Esterification With Cesium Salts. Helv. Chim. Acta. 56(5): 1476-1482.
      (2) Atherton E., Logan CJ, Sheppard RC. Peptide synthesis, Part 2. Procedures for solid-phase synthesis using N(alpha)-fluorenylmethoxycarbonylamino-acids on polyamide supports. Synthesis of substance P and of acyl carrier protein 65-74 decapeptide. J. Chem. Soc., Perkin Trans. 1. (0): 538-546.

      Helpful?

Reviews

No rating value

Active Filters