Skip to Content
Merck
All Photos(1)

Key Documents

443700

Sigma-Aldrich

Ethyl 4,4,4-trifluorobutyrate

≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€80.80

€80.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€80.80

About This Item

Linear Formula:
CF3CH2CH2CO2C2H5
CAS Number:
Molecular Weight:
170.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€80.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98%

form

liquid

refractive index

n20/D 1.3516 (lit.)

bp

127 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

functional group

ester
fluoro

SMILES string

CCOC(=O)CCC(F)(F)F

InChI

1S/C6H9F3O2/c1-2-11-5(10)3-4-6(7,8)9/h2-4H2,1H3

InChI key

PSRZMXNNQTWAGB-UHFFFAOYSA-N

Related Categories

General description

Ethyl 4,4,4-trifluorobutyrate is an ethyl ester of 4,4,4-tributyric acid. It is a useful spin trapping reagent used for ESR/EPR spectroscopy. It can be synthesized from Grignard reagent derived from 3-chloro-1,1,1-trifluoropropane..[1]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R T Jacobs et al.
Journal of medicinal chemistry, 37(9), 1282-1297 (1994-04-29)
The continued exploration of a series of 3-(arylmethyl)-1H-indole-5-carboxamides by the introduction of fluorinated amide substituents has resulted in the discovery of 4-[[5-[((2R)-2-methyl-4,4,4-trifluorobutyl)carbamoyl]-1-methyli ndol- 3-yl]methyl]-3-methoxy-N-[(2-methyl-phenyl)sulfonyl]benzamide (38p, ZENECA ZD3523), which has been chosen for clinical evaluation. This compound exhibited a Ki of
The Journal of Organic Chemistry, 60, 5434-5434 (1995)
Compounds Derived from 3-Halo-1, 1, 1-trifluoropropane1, 2.
McBee E, et al.
Journal of the American Chemical Society, 72(11), 5071-5073 (1950)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service