Skip to Content
Merck
All Photos(1)

Key Documents

257443

Sigma-Aldrich

Fluoroacetonitrile

98%

Synonym(s):

2-Fluoroacetonitrile, Fluoromethyl cyanide, Monofluoroacetonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€41.30

€41.30


Estimated to ship on30 March 2025


Request a Bulk Order

Select a Size

Change View
1 G
€41.30

About This Item

Linear Formula:
FCH2CN
CAS Number:
Molecular Weight:
59.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
98%

€41.30


Estimated to ship on30 March 2025


Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.333 (lit.)

bp

79-80 °C (lit.)

density

1.061 g/mL at 25 °C (lit.)

functional group

fluoro
nitrile

SMILES string

FCC#N

InChI

1S/C2H2FN/c3-1-2-4/h1H2

InChI key

GNFVFPBRMLIKIM-UHFFFAOYSA-N

Application

Fluoroacetonitrile has been used in preparation of:
  • 2-fluoromethyl-4,4,6-trimethyl-1,3-oxazine[1]
  • α-fluorinated acetophenone[2]
  • 2-amino-2-fluoromethyl-3-pentenenitrile, a key intermediate in the synthesis of 2,5-diamino-2-fluoromethyl-3(E)-pentenoic acid[3]

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

6.8 °F - closed cup

Flash Point(C)

-14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-Fluoromethyl-4, 4, 6-trimetiiyl-1, 3-oxasine as a new reagent for the preparation of a-fluoroaldehydes.
Patrick TB, et al.
Tetrahedron Letters, 31(2), 179-182 (1990)
Preparative flow techniques. 2. Grignard addition reaction on fluoroaceto-nitrile: Synthesis of 2-amino-2-fluoromethyl-3-pentenenitrile.
Gerhart F, et al.
Journal of Fluorine Chemistry, 50(2), 243-249 (1990)
Erum Raja et al.
Tetrahedron letters, 52(40), 5170-5172 (2012-03-03)
Fluoro-substituted aliphatic nitriles are shown to undergo the Houben-Hoesch reactions with arenes in CF(3)SO(3)H to give fluoro-substituted ketones in good yields. The fluorine substituents appear to enhance the reactivities of the nitriles (and the nitrilium ion intermediates) compared to similar
Santanu Mondal et al.
ACS chemical biology, 13(4), 1057-1065 (2018-03-09)
Protein arginine deiminases (PADs) play an important role in the pathogenesis of various diseases, including rheumatoid arthritis, multiple sclerosis, lupus, ulcerative colitis, and breast cancer. Therefore, the development of PAD inhibitors has drawn significant research interest in recent years. Herein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service