Skip to Content
Merck
All Photos(1)

Key Documents

269697

Sigma-Aldrich

Ethyl 4,4,4-trifluorocrotonate

98%

Synonym(s):

Ethyl 4,4,4-trifluoro-2-butenoate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€72.80

€72.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€72.80

About This Item

Linear Formula:
CF3CH=CHCO2C2H5
CAS Number:
Molecular Weight:
168.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€72.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.3601 (lit.)

bp

114-115 °C (lit.)

density

1.125 g/mL at 25 °C (lit.)

functional group

ester
fluoro

SMILES string

CCOC(=O)\C=C\C(F)(F)F

InChI

1S/C6H7F3O2/c1-2-11-5(10)3-4-6(7,8)9/h3-4H,2H2,1H3/b4-3+

InChI key

ZKRJCMKLCDWROR-ONEGZZNKSA-N

General description

The Michael reaction between ethyl 4,4,4-trifluorocrotonate and a Ni(II) complex of the Schiff base of glycine with (S)-o-[N-(N-benzylprolyl)amino]benzophenone was studied[1].

Application

Ethyl 4,4,4-trifluorocrotonate was used in the synthesis of (2S,3S)-3-methyl- and (2S,3S)-3-trifluoromethylpyroglutamic acid. It undergoes diastereoselective Michael addition reaction with ethyl crotonate[2].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient asymmetric synthesis of (2S,3S)-3-trifluoromethylpyroglutamic acid.
Tetrahedron Letters, 38(27), 4903-4904 (1997)
Stereochemically defined C-substituted glutamic acids and their derivatives. 1. An efficient asymmetric synthesis of (2S, 3S)-3-methyl-and-3-trifluoromethylpyroglutamic acids.
Soloshonok VA, et al.
Tetrahedron, 55(41), 12031-12044 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service