Skip to Content
Merck
All Photos(2)

Key Documents

436305

Sigma-Aldrich

Isobutyl acrylate

≥99%, contains 10-20 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

Isobutyl 2-propenoate, Isobutyl acrylate

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€96.10
1 L
€279.00

€96.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€96.10
1 L
€279.00

About This Item

Linear Formula:
CH2=CHCOOCH2CH(CH3)2
CAS Number:
Molecular Weight:
128.17
Beilstein:
1749388
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€96.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99%

form

liquid

contains

10-20 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.414 (lit.)

bp

132 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

SMILES string

CC(C)COC(=O)C=C

InChI

1S/C7H12O2/c1-4-7(8)9-5-6(2)3/h4,6H,1,5H2,2-3H3

InChI key

CFVWNXQPGQOHRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Design, control and economics of a process for isobutyl acrylate production: This study discusses the development of a process for producing isobutyl acrylate through the esterification of acrylic acid with isobutanol, emphasizing the challenges of separation processes involved (MD Moraru, DA Berinde, CS Bildea, 2018).
  • Liquid phase esterification of acrylic acid with isobutyl alcohol catalyzed by different cation exchange resins: Focuses on the esterification process to produce isobutyl acrylate using various catalysts, evaluating the efficiency and sustainability of the reaction (S Karakus, E Sert, AD Buluklu, 2014).
  • Synthesis of poly(isobutyl acrylate/n-butyl acrylate/methyl methacrylate)/CNC nanocomposites for adhesive applications via in situ semi-batch emulsion: Studies the synthesis of a nanocomposite material involving isobutyl acrylate for use in pressure-sensitive adhesives, highlighting the improved properties of the composite (A Ouzas, E Niinivaara, ED Cranston, 2019).

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatiana Smirnova et al.
Nanomaterials (Basel, Switzerland), 10(11) (2020-10-30)
Organic-inorganic photocurable nanocomposite materials are a topic of intensive research nowadays. The wide variety of materials and flexibility of their characteristics provide more freedom to design optical elements for light and neutron optics and holographic sensors. We propose a new
Jung-Hun Lee et al.
Polymers, 12(11) (2020-11-14)
Acrylic pressure-sensitive adhesives (PSAs) are used as fixatives between layers of a display. PSAs' function is an important factor that determines the performance of the display. Of the various display types available, the touch screen panel (TSP) of smart devices

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service