Skip to Content
Merck
All Photos(2)

Key Documents

E9706

Sigma-Aldrich

Ethyl acrylate

contains 10-20 ppm MEHQ as inhibitor, 99%

Synonym(s):

Acrylic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€42.40
1 L
€76.90
2 L
€93.90
10 L
€348.00

€42.40


Estimated to ship on29 March 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€42.40
1 L
€76.90
2 L
€93.90
10 L
€348.00

About This Item

Linear Formula:
CH2=CHCOOC2H5
CAS Number:
Molecular Weight:
100.12
Beilstein:
773866
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€42.40


Estimated to ship on29 March 2025


Request a Bulk Order

vapor density

3.5 (vs air)

Quality Level

vapor pressure

31 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

721 °F

contains

10-20 ppm MEHQ as inhibitor

expl. lim.

12.1 %

refractive index

n20/D 1.406 (lit.)

bp

99 °C (lit.)

mp

−71 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)C=C

InChI

1S/C5H8O2/c1-3-5(6)7-4-2/h3H,1,4H2,2H3

InChI key

JIGUQPWFLRLWPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

It is an acrylic monomer. It may be prepared either by reacting acetylene with carbon monoxide and ethyl alcohol in the presence of nickel carbonyl and hydrogen chloride as catalysts, or by catalyzed esterification of acrylic acid with alcohol. It polymerizes readily under the influence of heat, light or other initiators.

Application

Used in the manufacture of a variety of polymers and copolymers. It is also used as an ingredient in latex paints., binders , caulks, lubricating oils, polishes, adhesives, fibers, plastics and other products.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zahra Bazrafshan et al.
Nanomaterials (Basel, Switzerland), 8(9) (2018-09-13)
A collagen-based copolymer, ASC-g-Poly(methyl methacrylate-co-Ethyl Acrylate), was synthesized in the presence of Graphene Oxide (GO) via an in-situ polymerization. The presence of GO that increased the accessible surface area for initiated collagen chains allowed for an accelerated polymerization with highly
Zahra Bazrafshan et al.
Journal of the mechanical behavior of biomedical materials, 87, 19-29 (2018-07-22)
In this study, Acid Soluble Collagen-g-P(methyl methacrylate-co-ethyl acrylate) (CME) was synthesized to take advantage of the flexibility of the resulted branched polymer chains and the high density of their chain entanglement. The coaxial electrospinning technique was applied to study the
Andrea Venturato et al.
Macromolecular bioscience, 16(12), 1864-1872 (2016-10-26)
The development of polymeric materials with cell adhesion abilities requires an understanding of cell-surface interactions which vary with cell type. To investigate the correlation between cell attachment and the nature of the polymer, a series of random and block copolymers
Anna E V Hagström et al.
Biotechnology and bioengineering, 102(3), 693-699 (2008-09-30)
An OH-functional polyester has been acrylated via transesterification of ethyl acrylate, catalyzed by Candida antarctica lipase B (CalB) in two different preparations: Novozym 435 and immobilized on Accurel MP1000. The batch process resulted in incomplete acrylation as well as severe
M Monier et al.
Journal of hazardous materials, 176(1-3), 348-355 (2009-12-08)
The graft copolymerization of ethyl acrylate (EA) onto natural wool fibers initiated by potassium persulphate and Mohr's salt redox initiator system in limited aqueous medium was carried out in heterogeneous media. Ester groups of the grafted copolymers were partially converted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service