Skip to Content
Merck
All Photos(1)

Key Documents

909041

Sigma-Aldrich

Benzyl acrylate

≥99.8%, contains 40-60 ppm MEHQ as inhibitor

Synonym(s):

Acrylic acid benzyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€36.40

€36.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€36.40

About This Item

Empirical Formula (Hill Notation):
C10H10O2
CAS Number:
Molecular Weight:
162.19
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.23

€36.40


Check Cart for Availability

Request a Bulk Order

Assay

≥99.8%

form

liquid

contains

40-60 ppm MEHQ as inhibitor

color

colorless

SMILES string

O(Cc1ccccc1)C(=O)C=C

InChI

1S/C10H10O2/c1-2-10(11)12-8-9-6-4-3-5-7-9/h2-7H,1,8H2

InChI key

GCTPMLUUWLLESL-UHFFFAOYSA-N

Application

Benzyl acrylate is a high refractive index monomer. It can be used to prepare high refractive index polymers, and formulate inks.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

221.9 °F

Flash Point(C)

105.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongjin Seo et al.
Journal of colloid and interface science, 580, 776-784 (2020-07-28)
Many studies on the deformation of soft films by liquids confirmed the increase in the radius of the deformation and the decrease in the apparent contact angle. However, due to the thinness, the dynamics of the deformation could not be
Julia Kredel et al.
Polymers, 11(12) (2020-01-01)
Fluoropolymers represent a unique class of functional polymers due to their various interesting and important properties such as thermal stability, resistance toward chemicals, repellent behaviors, and their low refractive indices in comparison to other polymeric materials. Based on the latter

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service