Skip to Content
Merck
All Photos(1)

Key Documents

431184

Sigma-Aldrich

Poly(3,3′,4,4′-biphenyltetracarboxylic dianhydride-co-1,4-phenylenediamine), amic acid solution

9.5-11.5 wt. % in 1-methyl-2-pyrrolidinone, electronic grade

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€209.00

€209.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€209.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€209.00


Check Cart for Availability

Request a Bulk Order

grade

electronic grade

Quality Level

purified by

filtration through 1.0μm medium

concentration

9.5-11.5 wt. % in 1-methyl-2-pyrrolidinone

impurities

≤10 ppm total metals

refractive index

n20/D 1.497

viscosity

25-30 poise(25 °C, Brookfield)(lit.)

bp

202 °C

transition temp

Tg >400 °C (imide)

density

1.07 g/mL at 25 °C

anion traces

chloride (Cl-): ≤5 ppm

storage temp.

−20°C

SMILES string

Nc5ccc(cc5)N.O1C(=O)c2c(ccc(c2)c3cc4c(cc3)C(=O)OC4=O)C1=O

InChI

1S/C16H6O6.C6H8N2/c17-13-9-3-1-7(5-11(9)15(19)21-13)8-2-4-10-12(6-8)16(20)22-14(10)18;7-5-1-2-6(8)4-3-5/h1-6H;1-4H,7-8H2

InChI key

MUHWTMKSDCXLLV-UHFFFAOYSA-N

Application

Semi-conductor material. Dielectric component in thin films, high density interconnect systems and multichip modules.

Features and Benefits

High purity material with controlled metallic content. Polyimide has good thermal, mechanical and electrical properties.

Other Notes

Store at -18° C for extended shelf life.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service