Skip to Content
Merck
All Photos(2)

Key Documents

431176

Sigma-Aldrich

Poly(pyromellitic dianhydride-co-4,4′-oxydianiline), amic acid solution

electronic grade

Synonym(s):

Polyamic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€150.00

€150.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€150.00

About This Item

Linear Formula:
(C12H12N2O .C10H2O6)x
UNSPSC Code:
12162002
NACRES:
NA.23

€150.00


Check Cart for Availability

Request a Bulk Order

grade

electronic grade

Quality Level

purified by

sub-micron filtration (0.2 μm medium)

refractive index

n20/D 1.506

viscosity

30-38 poise(lit.)

bp

174 °C

transition temp

Tg >400 °C (imide)

density

1.04 g/mL at 25 °C

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Semi-conductor material. Dielectric component in thin films, high density interconnect systems and multichip modules.

Features and Benefits

Filtered through 0.2μm medium
High purity material with controlled metallic content. Polyimide has good thermal, mechanical and electrical properties.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 3 - Muta. 1B - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service