Skip to Content
Merck
All Photos(1)

Key Documents

524492

Sigma-Aldrich

4,4′-Oxydiphthalic anhydride

97%

Synonym(s):

3,3′,4,4′-Diphenyl ether tetracarboxylic acid dianhydride, 3,3′,4,4′-Diphenyl ether tetracarboxylic dianhydride, 3,3′,4,4′-Diphenyl oxide tetracarboxylic dianhydride, 5,5′-Oxybis[1,3-isobenzofurandione]

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€207.00

€207.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€207.00

About This Item

Empirical Formula (Hill Notation):
C16H6O7
CAS Number:
Molecular Weight:
310.21
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€207.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

225-229 °C (lit.)

SMILES string

O=C1OC(=O)c2cc(Oc3ccc4C(=O)OC(=O)c4c3)ccc12

InChI

1S/C16H6O7/c17-13-9-3-1-7(5-11(9)15(19)22-13)21-8-2-4-10-12(6-8)16(20)23-14(10)18/h1-6H

InChI key

QQGYZOYWNCKGEK-UHFFFAOYSA-N

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng-Yang Huang et al.
Nanomaterials (Basel, Switzerland), 10(1) (2020-01-02)
This study investigated films with remarkably high electrical conductivity after they were easily prepared from organic/inorganic nanohybrid solutions containing an organic polymeric dispersant and two-dimensional nanoscale silicate platelets as the inorganic stabilizer dispersed with silver nanoparticles. Transmission electron microscopy shows
Meng-Ge Huangfu et al.
Polymers, 11(6) (2019-06-05)
Protection of intrinsically brittle quartz chromatographic columns (CCs) from breakage or property deterioration in gas chromatography (GC) analysis has become an important research topic regarding high-temperature GC techniques. Polyimide (PI) has proved to be the most suitable protective coating for
An Yang et al.
Dental materials journal, 36(5), 560-565 (2017-06-02)
The aim of the study was to evaluate the effect of modifying polymethyl methacrylate (PMMA) denture base material with polyimide (PI) on its flexural property and biocompatibility. Low molecular weight (1,500 g/mol) PI was synthesized and small amount of PI

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service