Skip to Content
Merck
All Photos(1)

Key Documents

43081

Sigma-Aldrich

4-(Diphenylhydroxymethyl)benzoic acid

≥98.0% (HPLC)

Synonym(s):

4-(α-Hydroxybenzhydryl)benzoic acid, α,α-Diphenyl-α-hydroxy-p-toluic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€165.00

€165.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€165.00

About This Item

Empirical Formula (Hill Notation):
C20H16O3
CAS Number:
Molecular Weight:
304.34
Beilstein:
2660895
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

€165.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

solid

reaction suitability

reagent type: cross-linking reagent

mp

210-215 °C

functional group

carboxylic acid
hydroxyl

SMILES string

OC(C1=CC=CC=C1)(C2=CC=C(C(O)=O)C=C2)C3=CC=CC=C3

InChI

1S/C20H16O3/c21-19(22)15-11-13-18(14-12-15)20(23,16-7-3-1-4-8-16)17-9-5-2-6-10-17/h1-14,23H,(H,21,22)

InChI key

CPZWBWPALJLUBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Precursor of a new trityl linker. It is ideally suited for coupling with TentaGel[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B. Henkel et al.
Liebigs Ann./Recueil, 2161-2161 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service