Skip to Content
Merck

Skip To

8.55133

Fmoc-PAL AM resin

Novabiochem®

Synonym(s):

Fmoc-protected PAL AM bead

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
€ 95,50
5 g
Check Cart for Availability
€ 381,00

About This Item

NACRES:
NA.22
UNSPSC Code:
12352209

€ 95,50


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc, amine

storage temp.

2-8°C

General description

Fmoc-PAL-AM resin consists of Barany′s aminomethyl-dimethoxyphenoxyvaleric acid linker [1] attached to aminomethylated polystyrene. The amino group of this linker can be easily acylated under standard coupling conditions. Following peptide assembly, treatment with 95% TFA containing scavengers releases the desired peptide amide. Studies have shown the acid sensitivity of this linker to be around twice that of the Rink amide linker [2]. There is some evidence to suggest that PAL resins give greater yields in microwave assisted synthesis.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Analysis Note

Colour (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.50 - 0.80 mmol/g
Swelling Volume (in DMF): lot specific result

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.551378.550018.55134
reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

functional group

Fmoc, amine

functional group

amine

functional group

amine

functional group

amine

product line

Novabiochem®

product line

NovaSyn® TG, Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

Quality Level

200

Quality Level

200

Quality Level

400

Quality Level

200

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

form

beads

form

beads

form

beads

form

beads


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters