Skip to Content
Merck
All Photos(1)

Documents

369616

Supelco

Tetrahexylammonium hydrogensulfate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)5]4N(HSO4)
CAS Number:
Molecular Weight:
451.75
Beilstein:
4629729
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

Assay

98%

mp

98-100 °C (lit.)

SMILES string

OS([O-])(=O)=O.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

InChI

1S/C24H52N.H2O4S/c1-5-9-13-17-21-25(22-18-14-10-6-2,23-19-15-11-7-3)24-20-16-12-8-4;1-5(2,3)4/h5-24H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

RULHPTADXJPDSN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeynep S Teksin et al.
Journal of controlled release : official journal of the Controlled Release Society, 116(1), 50-57 (2006-10-20)
Parallel Artificial Membrane Permeability Assay (PAMPA) is a method to screen drug candidates for membrane permeability. The objective was to characterize the transport of a model weak base, metoprolol, across a three lipid-component PAMPA system (denoted A-PAMPA, for anionic-PAMPA) and
The effects of tetrahexylammonium chloride on calcium mobilization from cerebellar microsomes.
L G Sayers et al.
Biochemical Society transactions, 21(2), 105S-105S (1993-05-01)
N Surendran et al.
Journal of chromatography. B, Biomedical sciences and applications, 691(2), 305-312 (1997-04-11)
The objective of this research was to develop a rapid, sensitive and reliable method for the separation of phosphonodipeptide prodrugs and parent compounds to facilitate the evaluation of cell permeation using in vitro cell culture models. Separation was accomplished isocratically
Xuemei Wang et al.
Journal of biomedical materials research. Part A, 80(4), 852-860 (2006-10-31)
Although much effort has been extended to the efficient cancer therapies, the drug resistance is still a major obstacle in cancer chemotherapeutic treatments. Almost 90% of the cancer therapy failure is caused by the relative problems. Recently, the application of
A M Lisi et al.
Journal of chromatography, 581(1), 57-63 (1992-10-02)
A simple and efficient procedure has been developed for the derivatization of diuretic agents in human urine by direct extractive alkylation and their detection by gas chromatography-mass spectrometry. The procedure is an improvement over previous extractive alkylation methods because of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service