Skip to Content
Merck
All Photos(1)

Documents

87726

Sigma-Aldrich

Tetramethylammonium hydrogen sulfate monohydrate

crystallized, ≥98.0% (T)

Synonym(s):

N,N,N-trimethyl-methanaminium sulfate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(HSO4) · H2O
CAS Number:
Molecular Weight:
189.23
Beilstein:
4829202
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (T)

form

crystals

quality

crystallized

SMILES string

O.C[N+](C)(C)C.OS([O-])(=O)=O

InChI

1S/C4H12N.H2O4S.H2O/c2*1-5(2,3)4;/h1-4H3;(H2,1,2,3,4);1H2/q+1;;/p-1

InChI key

VSJXNAHOWBCQRE-UHFFFAOYSA-M

General description

Crystals of tetramethylammonium hydrogen sulfate monohydrate generally exhibit ferroelectricity in the temperature range from -104 °C to 40 °C.

Application

Tetramethylammonium hydrogen sulfate monohydrate can be used as a phase transfer catalyst in polymer synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ferroelectric properties of tetramethylammonium hydrogen sulfate hydrate (N(CH3)?4HSO4.H2O).
Suzuki, Haruhiko et al.
Jpn J. App. Phys., Part I, 24, 353-355 (1985)
Spontaneous polymer thin film assembly and organization using mutually immiscible side chains.
Sun, F et al.
Journal of the American Chemical Society, 118(8), 1856-1866 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service