Skip to Content
Merck
All Photos(1)

Key Documents

87299

Supelco

Tetrahexylammonium hydrogensulfate

suitable for ion pair chromatography, LiChropur, ≥99.0% (T)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€238.00
25 G
€1,460.00

€238.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€238.00
25 G
€1,460.00

About This Item

Linear Formula:
[CH3(CH2)5]4N(HSO4)
CAS Number:
Molecular Weight:
451.75
Beilstein:
4629729
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

€238.00


Estimated to ship on31 May 2025


Request a Bulk Order

description

cationic

Quality Level

Assay

≥99.0% (T)

form

crystals

quality

LiChropur

technique(s)

ion pair chromatography: suitable

mp

98-100 °C (lit.)
99-102 °C

λ

10 % in acetonitrile

UV absorption

λ: 210 nm Amax: 0.10
λ: 220 nm Amax: 0.04
λ: 230 nm Amax: 0.03
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

OS([O-])(=O)=O.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

InChI

1S/C24H52N.H2O4S/c1-5-9-13-17-21-25(22-18-14-10-6-2,23-19-15-11-7-3)24-20-16-12-8-4;1-5(2,3)4/h5-24H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

RULHPTADXJPDSN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahexylammonium hydrogensulfate is a quaternary ammonium salt mainly used as an electrolyte.

Application

Tetrahexylammonium hydrogensulfate may have been used as a phase-transfer catalyst during catalysis analysis of benzylic halide to carboxylic acid.[1]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective phase transfer and palladium (0)-catalyzed carbonylation, carbalkoxylation, and reduction reactions.
Alper, Howard, Khaled Hashem, and Josef Heveling.
Organometallics, 1.6, 775-778 (1982)
A M Lisi et al.
Journal of chromatography, 581(1), 57-63 (1992-10-02)
A simple and efficient procedure has been developed for the derivatization of diuretic agents in human urine by direct extractive alkylation and their detection by gas chromatography-mass spectrometry. The procedure is an improvement over previous extractive alkylation methods because of
N Surendran et al.
Journal of chromatography. B, Biomedical sciences and applications, 691(2), 305-312 (1997-04-11)
The objective of this research was to develop a rapid, sensitive and reliable method for the separation of phosphonodipeptide prodrugs and parent compounds to facilitate the evaluation of cell permeation using in vitro cell culture models. Separation was accomplished isocratically
Xuemei Wang et al.
Journal of biomedical materials research. Part A, 80(4), 852-860 (2006-10-31)
Although much effort has been extended to the efficient cancer therapies, the drug resistance is still a major obstacle in cancer chemotherapeutic treatments. Almost 90% of the cancer therapy failure is caused by the relative problems. Recently, the application of
Y Uyama et al.
Pflugers Archiv : European journal of physiology, 426(5), 363-370 (1994-03-01)
Effects of tetraalkylammonium ions, having tetraalkyl chains of increasing length from ethyl to octyl, on inositol-trisphosphate (InsP3)-induced Ca2+ release and contractile mechanics were examined in guinea-pig skinned ileal smooth muscle longitudinal strips. Although tetrahexylammonium ions (THexA) appeared to be the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service