Skip to Content
Merck
All Photos(1)

Key Documents

367931

Sigma-Aldrich

2-Bromo-4-fluorobenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(F)CO2H
CAS Number:
Molecular Weight:
219.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

172-176 °C (lit.)

SMILES string

OC(=O)c1ccc(F)cc1Br

InChI

1S/C7H4BrFO2/c8-6-3-4(9)1-2-5(6)7(10)11/h1-3H,(H,10,11)

InChI key

RRKPMLZRLKTDQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amination of 2-bromo-4-fluorobenzoic acid with aniline is reported to yield N-phenyl-4-fluoro-anthranilic acid.[1]

Application

2-Bromo-4-fluorobenzoic acid may be used in the synthesis of:
  • 3-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one[2]
  • 2-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one[2]
  • 2-((2-carboxy-5-fluorophenyl)amino)-3-methoxybenzoic acid[3]
  • 2-bromo-4-fluorobenzamide[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuroleptics with protracted action: 3-Fluoro derivatives of methiothepin and oxyprothepin and their 2-fluoro analogues.
Kopicova Z, et al.
Collection of Czechoslovak Chemical Communications, 40(11), 3519-3529 (1975)
Bernd Dörner et al.
Bioorganic & medicinal chemistry, 19(7), 2190-2198 (2011-03-23)
Aim of this study was to label the potent dual P-glycoprotein (Pgp) and breast cancer resistance protein (BCRP) inhibitor elacridar (1) with (18)F to provide a positron emission tomography (PET) radiotracer to visualize Pgp and BCRP. A series of new
Lauren R Donaldson et al.
Organic & biomolecular chemistry, 9(7), 2233-2239 (2011-02-08)
A Heck cyclisation approach is described for the rapid synthesis of a library of natural product-like small molecules, based on the phenanthridine core. The synthesis of a range of substituted benzylamine building blocks and their incorporation into the library is
Christian Wolf et al.
The Journal of organic chemistry, 71(8), 3270-3273 (2006-04-08)
A chemo- and regioselective copper-catalyzed cross-coupling procedure for amination of 2-bromobenzoic acids is described. The method eliminates the need for acid protection and produces N-aryl and N-alkyl anthranilic acid derivatives in up to 99% yield. N-(1-Pyrene)anthranilic acid has been employed
Fu-Xiang Wang et al.
Developmental cell, 54(6), 742-757 (2020-08-07)
Plant somatic embryogenesis refers to a phenomenon where embryos develop from somatic cells in the absence of fertilization. Previous studies have revealed that the phytohormone auxin plays a crucial role in somatic embryogenesis by inducing a cell totipotent state, although

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service