Skip to Content
Merck
All Photos(1)

Key Documents

392383

Sigma-Aldrich

4-Bromo-2-fluorobenzoic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€64.80

€64.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€64.80

About This Item

Linear Formula:
BrC6H3(F)COOH
CAS Number:
Molecular Weight:
219.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€64.80


Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

211-215 °C (lit.)

functional group

bromo
carboxylic acid
fluoro

SMILES string

OC(=O)c1ccc(Br)cc1F

InChI

1S/C7H4BrFO2/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3H,(H,10,11)

InChI key

ZQQSRVPOAHYHEL-UHFFFAOYSA-N

General description

4-Bromo-2-fluorobenzoic acid is a halogen substituted benzoic acid.

Application

4-Bromo-2-fluorobenzoic acid may be used in the synthesis of biaryl intermediates by palladium-mediated coupling with various aryl boronic acids.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Yong Choi et al.
ACS medicinal chemistry letters, 5(4), 434-439 (2014-06-06)
Sterol 14α-demethylase (CYP51) is an important therapeutic target for fungal and parasitic infections due to its key role in the biosynthesis of ergosterol, an essential component of the cell membranes of these pathogenic organisms. We report the development of potent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service