Skip to Content
Merck
All Photos(1)

Documents

290203

Sigma-Aldrich

3,5-Difluorobenzonitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CN
CAS Number:
Molecular Weight:
139.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

84-86 °C (lit.)

SMILES string

Fc1cc(F)cc(c1)C#N

InChI

1S/C7H3F2N/c8-6-1-5(4-10)2-7(9)3-6/h1-3H

InChI key

CQXZSEXZQVKCHW-UHFFFAOYSA-N

General description

The vibrational wavenumbers, geometry and various thermodynamic parameters were studied for 3,5-difluorobenzonitrile.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V K Rastogi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 58(9), 1987-2004 (2002-08-08)
Geometry, vibrational wavenumbers and several thermodynamic parameters were calculated using ab initio quantum chemical methods for the 3,5-difluorobenzonitrile molecule. The results were compared with the experimental values. With the help of three specific scaling procedures, the observed vibrational wavenumbers in
Mounggon Kim et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(7), 1829-1834 (2018-11-27)
Molecular design principles of thermally activated delayed fluorescent (TADF) emitters having a high quantum efficiency and a color tuning capability was investigated by synthesizing three TADF emitters with donors at different positions of a benzonitrile acceptor. The position rendering a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service