Skip to Content
Merck
All Photos(1)

Key Documents

360023

Sigma-Aldrich

o-Tolylmagnesium chloride solution

1.0 M in THF

Synonym(s):

2-Methylphenylmagnesium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€89.80
800 ML
€374.00

€89.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€89.80
800 ML
€374.00

About This Item

Linear Formula:
CH3C6H4MgCl
CAS Number:
Molecular Weight:
150.89
Beilstein:
3537703
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€89.80


Check Cart for Availability

Request a Bulk Order

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in THF

density

0.956 g/mL at 25 °C

SMILES string

Cc1ccccc1[Mg]Cl

InChI

1S/C7H7.ClH.Mg/c1-7-5-3-2-4-6-7;;/h2-5H,1H3;1H;/q;;+1/p-1

InChI key

DSJSQDNPAOUFOJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

3.2 °F - closed cup

Flash Point(C)

-16 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oleg V Larionov et al.
Organic letters, 16(3), 864-867 (2014-01-15)
A one-step transformation of heterocyclic N-oxides to 2-alkyl-, aryl-, and alkenyl-substituted N-heterocycles is described. The success of this broad-scope methodology hinges on the combination of copper catalysis and activation by lithium fluoride or magnesium chloride. The utility of this method

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service