Skip to Content
Merck
All Photos(1)

Key Documents

224499

Sigma-Aldrich

tert-Butylmagnesium chloride solution

2.0 M in diethyl ether

Synonym(s):

t-Butylmagnesium chloride, tert-Butylchloromagnesium, Chloro(1,1-dimethylethyl)magnesium

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€74.70
800 ML
€359.00

€74.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€74.70
800 ML
€359.00

About This Item

Linear Formula:
(CH3)3CMgCl
CAS Number:
Molecular Weight:
116.87
Beilstein:
3535403
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€74.70


Check Cart for Availability

Request a Bulk Order

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

2.0 M in diethyl ether

density

0.828 g/mL at 25 °C

SMILES string

CC(C)(C)[Mg]Cl

InChI

1S/C4H9.ClH.Mg/c1-4(2)3;;/h1-3H3;1H;/q;;+1/p-1

InChI key

ZDRJSYVHDMFHSC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

tert-Butylmagnesium chloride solution can be used as a Grignard reagent:
  • In the Fe-catalyzed cross-coupling reaction with chloroenynols[1].
  • To synthesize alkyne-terminated polyhexylthiophenes[2].

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self-assembly of carbohydrate-block-poly (3-hexylthiophene) diblock copolymers into sub-10 nm scale lamellar structures
Sakai-Otsuka Y, et al.
Macromolecules, 50, 3365-3376 (2017)
Abdallah Hamze et al.
The Journal of organic chemistry, 72(10), 3868-3874 (2007-04-17)
Palladium-catalyzed hydrostannation of substituted Z- and E-enynols is discussed and compared. The regioselectivity of the H-Sn bond addition was found to be controlled by the geometry of the double bond (Z- or syn-directing effect) rather than the nature of its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service