Skip to Content
Merck
All Photos(2)

Key Documents

C24795

Sigma-Aldrich

2-Chlorobenzonitrile

98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€64.20

€64.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€64.20

About This Item

Linear Formula:
ClC6H4CN
CAS Number:
Molecular Weight:
137.57
Beilstein:
1363916
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€64.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

crystals

bp

232 °C (lit.)

mp

43-46 °C (lit.)

SMILES string

Clc1ccccc1C#N

InChI

1S/C7H4ClN/c8-7-4-2-1-3-6(7)5-9/h1-4H

InChI key

NHWQMJMIYICNBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leandro Val Sayson et al.
Psychopharmacology, 236(7), 2201-2210 (2019-03-21)
Depressive syndrome or depression is a debilitating brain disorder affecting numerous people worldwide. Although readily available, current antidepressants have low remission rates and late onset times. Recently, N-methyl-D-aspartate (NMDA) receptor antagonists, like ketamine and methoxetamine (MXE), were found to elicit

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service