Skip to Content
Merck
All Photos(1)

Documents

348112

Sigma-Aldrich

Triethylvinylsilane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHSi(C2H5)3
CAS Number:
Molecular Weight:
142.31
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.434 (lit.)

bp

146-147 °C (lit.)

density

0.771 g/mL at 25 °C (lit.)

SMILES string

CC[Si](CC)(CC)C=C

InChI

1S/C8H18Si/c1-5-9(6-2,7-3)8-4/h5H,1,6-8H2,2-4H3

InChI key

HBWGDHDXAMFADB-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service