Skip to Content
Merck
All Photos(2)

Key Documents

446203

Sigma-Aldrich

Dimethoxymethylvinylsilane

97%

Synonym(s):

Dimethoxymethylvinylsilane, Methyldimethoxyvinylsilane, Vinyldimethoxymethylsilane

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€117.00
100 ML
€376.00

€117.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€117.00
100 ML
€376.00

About This Item

Linear Formula:
CH3Si(OCH3)2CH=CH2
CAS Number:
Molecular Weight:
132.23
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€117.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.395 (lit.)

bp

106 °C (lit.)

density

0.884 g/mL at 25 °C (lit.)

SMILES string

CO[Si](C)(OC)C=C

InChI

1S/C5H12O2Si/c1-5-8(4,6-2)7-3/h5H,1H2,2-4H3

InChI key

ZLNAFSPCNATQPQ-UHFFFAOYSA-N

Application

Dimethoxymethylvinylsilane may be used in the synthesis of polymethylvinylborosiloxanes (PMVBs) by direct polycondensation.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

37.4 °F - closed cup

Flash Point(C)

3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alexander N Bugrov et al.
Luminescence : the journal of biological and chemical luminescence, 33(5), 837-849 (2018-04-24)
Spherical nanoparticles of ZrO2 with 2 and 10 mol% EuO1.5 up to 20 nm size were prepared by the method of hydrothermal synthesis for luminescent functionalization of the polymer-inorganic nanocomposites based on poly(methyl methacrylate). Surface modification of oxide nanoparticles was carried out
Synthesis of hyperbranched polymethylvinylborosiloxanes and modification of addition-curable silicone with improved thermal stability.
Li L, et al.
Applied Organometallic Chemistry, 27(12), 723-728 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service