Skip to Content
Merck
All Photos(1)

Key Documents

347558

Sigma-Aldrich

4-Oxo-2-thioxo-3-thiazolidinylacetic acid

≥99%

Synonym(s):

Rhodanine-3-acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€39.30

€39.30


Estimated to ship on17 April 2025


Request a Bulk Order

Select a Size

Change View
5 G
€39.30

About This Item

Empirical Formula (Hill Notation):
C5H5NO3S2
CAS Number:
Molecular Weight:
191.23
Beilstein:
149513
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€39.30


Estimated to ship on17 April 2025


Request a Bulk Order

Quality Level

Assay

≥99%

form

solid

mp

145-148 °C (lit.)

solubility

methanol: soluble 25 mg/mL, clear, yellow

functional group

carboxylic acid
thioether

SMILES string

OC(=O)CN1C(=O)CSC1=S

InChI

1S/C5H5NO3S2/c7-3-2-11-5(10)6(3)1-4(8)9/h1-2H2,(H,8,9)

InChI key

JGRMXPSUZIYDRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Oxo-2-thioxo-3-thiazolidinylacetic acid was employed as sample to investigate the uncertainty in organic elemental analysis of C, H, N and S.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Companyó et al.
Analytical and bioanalytical chemistry, 392(7-8), 1497-1505 (2008-10-15)
We evaluate the uncertainty in organic elemental analysis of C, H, N, and S. We use data from six proficiency tests (PTs), in which some 35 Spanish laboratories participated. The uncertainty of the technique is estimated from the relative within-laboratory
Zhen-Hua Chen et al.
European journal of medicinal chemistry, 45(12), 5739-5743 (2010-10-05)
With an intention to synergize the anti-bacterial activity of chalcones and rhodanine-3-acetic acid, several hybrid compounds possessing chalcone and rhodanine-3-acetic acid moieties were synthesized and tested for their anti-bacterial activity. Some compounds presented great anti-microbial activities against Gram-positive bacteria (including
Ana Martinez et al.
Journal of medicinal chemistry, 48(23), 7103-7112 (2005-11-11)
The 2,4-disubstituted thiadiazolidinones (TDZD) are described as the first ATP-noncompetitive GSK-3 inhibitors. Following an SAR study about TDZD, different structural modifications in the heterocyclic ring aimed to test the influence of each heteroatom on the biological study are here reported

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service