Skip to Content
Merck
All Photos(1)

Key Documents

112275

Sigma-Aldrich

α-Methyl-trans-cinnamaldehyde

98%

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€43.00

€43.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€43.00

About This Item

Linear Formula:
C6H5CH=C(CH3)CHO
CAS Number:
Molecular Weight:
146.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€43.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

refractive index

n20/D 1.605 (lit.)

bp

148-149 °C/27 mmHg (lit.)

density

1.047 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)\C(C)=C(/[H])c1ccccc1

InChI

1S/C10H10O/c1-9(8-11)7-10-5-3-2-4-6-10/h2-8H,1H3/b9-7+

InChI key

VLUMOWNVWOXZAU-VQHVLOKHSA-N

Biochem/physiol Actions

α-Methyl trans cinnamaldehyde has antifungal activity. It is self coupled and complexed with Co(II) and Ni(II) to synthesize ligand and complexes[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheikh Shreaz et al.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 24(5), 923-933 (2011-04-09)
Antifungal effectivity and utility of cinnamaldehyde is limited because of its high MIC and skin sensitivity. In this study, α-methyl trans cinnamaldehyde, a less irritating derivative, have been self coupled and complexed with Co(II) and Ni(II) to generate N, N'-Bis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service