Skip to Content
Merck
All Photos(1)

Key Documents

A79507

Sigma-Aldrich

3-Aminorhodanine

99%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€72.10

€72.10


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
10 G
€72.10

About This Item

Empirical Formula (Hill Notation):
C3H4N2OS2
CAS Number:
Molecular Weight:
148.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€72.10


Estimated to ship on14 April 2025


Request a Bulk Order

Assay

99%

form

solid

mp

100-103 °C (lit.)

SMILES string

NN1C(=O)CSC1=S

InChI

1S/C3H4N2OS2/c4-5-2(6)1-8-3(5)7/h1,4H2

InChI key

ZBUUHLDYMKTVLT-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N E Madadi et al.
Bollettino chimico farmaceutico, 130(4), 124-127 (1991-04-01)
Two series of hydrazones were prepared by condensation of various aldehydes with 2-aminorhodanine (series 1) and 2-hydrazino-2-imidazoline (series 2). These compounds showed interesting antiparasitic activities against amoeba, trichomonas, leishmania and plasmodium.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service