Skip to Content
Merck
All Photos(1)

Documents

340723

Sigma-Aldrich

2,6-Dibromo-4-fluoroaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C6H2(F)NH2
CAS Number:
Molecular Weight:
268.91
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

64-66 °C (lit.)

SMILES string

Nc1c(Br)cc(F)cc1Br

InChI

1S/C6H4Br2FN/c7-4-1-3(9)2-5(8)6(4)10/h1-2H,10H2

InChI key

VHPLZFGCNLDYQH-UHFFFAOYSA-N

Application

2,6-Dibromo-4-fluoroaniline was employed in the asymmetric synthesis of prostaglandin D2 receptor antagonist for the treatment of allergic rhinitis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin R Campos et al.
The Journal of organic chemistry, 70(1), 268-274 (2004-12-31)
An asymmetric synthesis was developed for the production of a prostaglandin D(2) receptor antagonist for the treatment of allergic rhinitis. The stereogenic center was set using asymmetric allylic alkylation chemistry, and the core of the structure was constructed via Pd-catalyzed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service