Skip to Content
Merck
All Photos(1)

Key Documents

233544

Sigma-Aldrich

4-Dodecylaniline

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€176.00

€176.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€176.00

About This Item

Linear Formula:
CH3(CH2)11C6H4NH2
CAS Number:
Molecular Weight:
261.45
Beilstein:
2646305
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€176.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

bp

220-221 °C/15 mmHg (lit.)

mp

35-39 °C (lit.)

SMILES string

CCCCCCCCCCCCc1ccc(N)cc1

InChI

1S/C18H31N/c1-2-3-4-5-6-7-8-9-10-11-12-17-13-15-18(19)16-14-17/h13-16H,2-12,19H2,1H3

InChI key

KLPPPIIIEMUEGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Dodecylaniline has been used:
  • in the synthesis of sodium 4-dodecylphenylazosulfonate[1]
  • to investigate the water solubilization of single-walled carbon nanotubes (SWNTs)[2]
  • in proton transfer step during molecular oxygen (O2) reduction by decamethylferrocene (DMFc)[3]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su B, et al.
Oxygen reduction by decamethylferrocene at liquid/liquid interfaces catalyzed by dodecylaniline.
Journal of Electroanalytical Chemistry, 639(1), 102-108 (2010)
Water solubilization of single-walled carbon nanotubes by functionalization with glucosamine.
Pompeo F and Resasco DE.
Nano Letters, 2(4), 369-373 (2002)
A photodestructible surfactant.
Dunkin IR, et al.
Journal of the Chemical Society. Chemical Communications, 19, 2245-2246 (1994)
Kyle A Brown et al.
Nature methods, 16(5), 417-420 (2019-04-17)
We report the identification of a photocleavable anionic surfactant, 4-hexylphenylazosulfonate (Azo), which can be rapidly degraded by ultraviolet irradiation, for top-down proteomics. Azo can effectively solubilize proteins with performance comparable to that of sodium dodecyl sulfate (SDS) and is compatible

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service