Skip to Content
Merck
All Photos(1)

Key Documents

304352

Sigma-Aldrich

2,3,4-Trifluoroaniline

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€25.60

€25.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€25.60

About This Item

Linear Formula:
F3C6H2NH2
CAS Number:
Molecular Weight:
147.10
Beilstein:
3245609
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€25.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.486 (lit.)

bp

92 °C/48 mmHg (lit.)

density

1.393 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Nc1ccc(F)c(F)c1F

InChI

1S/C6H4F3N/c7-3-1-2-4(10)6(9)5(3)8/h1-2H,10H2

InChI key

WRDGNXCXTDDYBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Study on synthesis of 2,3,4-trifluoroaniline was reported.[1]

Application

2,3,4-Trifluoroaniline was used in preparation of diethyl 2-(2,3,4-trifluoro)-phenylaminomethylene malonate.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study on Synthesis of 2, 3, 4-Trifluoroaniline.
Tai-biaoZ, et al.
Fine Chemicals / ????, Z1-Z1 (2007)
Thermochemical reaction of 7-azido-1-ethyl-6, 8-difluoroquinolone-3-carboxylate with heterocyclic amines. An expeditious synthesis of novel fluoroquinolone derivatives.
Leyva S and Leyva E.
Tetrahedron, 63(9), 2093-2097 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service