Skip to Content
Merck
All Photos(1)

Key Documents

336971

Sigma-Aldrich

Tripropylphosphine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)3P
CAS Number:
Molecular Weight:
160.24
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.4584 (lit.)

bp

72-74 °C/12 mmHg (lit.)

density

0.801 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

CCCP(CCC)CCC

InChI

1S/C9H21P/c1-4-7-10(8-5-2)9-6-3/h4-9H2,1-3H3

InChI key

KCTAHLRCZMOTKM-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johannes Wiest et al.
Journal of controlled release : official journal of the Controlled Release Society, 268, 314-322 (2017-11-04)
Poor water solubility of drugs fuels complex formulations and jeopardizes patient access to medication. Simplifying these complexities we systematically synthesized a library of 36 sterically demanding counterions and mapped the pharmaceutical design space for amorphous ionic liquid strategies for Selurampanel
Rahul A Patil et al.
Analytical and bioanalytical chemistry, 410(19), 4645-4655 (2018-01-31)
The rapid expansion in the study and use of ionic liquids (ILs) is a result of their unique properties including negligible volatility, high thermal stability, and ability to dissolve disparate compounds. However, because ILs have infinitely variable structures (often referred

Related Content

The Catalexis platform enhances catalysis by digitally optimizing catalyst selection to identify the most effective phosphine ligands for cross-coupling reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service