Skip to Content
Merck
All Photos(1)

Key Documents

323314

Sigma-Aldrich

Tetrabutylammonium thiocyanate

98%

Synonym(s):

Tetrabutylammonium rhodanide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€79.00

€79.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€79.00

About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(SCN)
CAS Number:
Molecular Weight:
300.55
Beilstein:
3919896
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€79.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

120-123 °C (lit.)

SMILES string

[S-]C#N.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.CHNS/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;2-1-3/h5-16H2,1-4H3;3H/q+1;/p-1

InChI key

RSHBFZCIFFBTEW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michelle L Ingalsbe et al.
Bioorganic & medicinal chemistry letters, 19(17), 4984-4987 (2009-08-08)
Due to the increasing number of strains of drug-resistant bacteria, the development of new antibiotics has become increasingly important. The antibacterial properties of quaternary amines and their derivatives on both Gram-positive and Gram-negative bacteria are well known. However, an encompassing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service