Skip to Content
Merck
All Photos(2)

Key Documents

299669

Sigma-Aldrich

Silver thiocyanate

99%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€94.90

€94.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€94.90

About This Item

Linear Formula:
AgSCN
CAS Number:
Molecular Weight:
165.95
Beilstein:
3623095
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder
solubility:
H2O: insoluble(lit.)

€94.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

powder

solubility

H2O: insoluble(lit.)

SMILES string

[Ag]SC#N

InChI

1S/CHNS.Ag/c2-1-3;/h3H;/q;+1/p-1

InChI key

RHUVFRWZKMEWNS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service