Skip to Content
Merck
All Photos(1)

Key Documents

317160

Sigma-Aldrich

4-Chloro-2-fluoroaniline

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€67.90

€67.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€67.90

About This Item

Linear Formula:
ClC6H3(F)NH2
CAS Number:
Molecular Weight:
145.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€67.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.56 (lit.)

density

1.311 g/mL at 25 °C (lit.)

functional group

chloro
fluoro

SMILES string

Nc1ccc(Cl)cc1F

InChI

1S/C6H5ClFN/c7-4-1-2-6(9)5(8)3-4/h1-3H,9H2

InChI key

CSFDTBRRIBJILD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service