Skip to Content
Merck
All Photos(1)

Key Documents

196606

Sigma-Aldrich

2,5-Difluoroaniline

99.5%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€29.80

€29.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€29.80

About This Item

Linear Formula:
F2C6H3NH2
CAS Number:
Molecular Weight:
129.11
Beilstein:
2802549
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€29.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.5%

refractive index

n20/D 1.513 (lit.)

bp

176-178 °C (lit.)

mp

11-13 °C (lit.)

density

1.288 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Nc1cc(F)ccc1F

InChI

1S/C6H5F2N/c7-4-1-2-5(8)6(9)3-4/h1-3H,9H2

InChI key

YNOOQIUSYGWMSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,5-Difluoroaniline was used in the synthesis of 2,4-di-tert-butyl-6-[(2,5-difluorophenyl)iminomethyl]phenol[1]. It was also used in ultrasound-assisted preparation of 1,4-diazabutadienes[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jing-Yu He et al.
Ultrasonics sonochemistry, 18(1), 466-469 (2010-08-28)
An ultrasound-assisted preparation of 1,4-diazabutadienes via smooth condensation of diketones with amines under solvent-free conditions is described. The generality of this method was examined by the synthesized N,N'-diaryl- and N,N'-dialkyl-1,4-diazabutadiene derivatives. In addition to experimental simplicity, the main advantages of
Omer Celik et al.
Acta crystallographica. Section E, Structure reports online, 65(Pt 11), o2786-o2786 (2009-01-01)
In the title Schiff base, C(21)H(25)F(2)NO, the dihedral angle between the aromatic rings is 27.90(5)° and an intramolecular O-H⋯N hydrogen bond occurs. In the crystal, the molecules are linked by C-H⋯O, C-H⋯N and C-H⋯F interactions.
I M Rietjens et al.
Chemico-biological interactions, 94(1), 49-72 (1995-01-01)
The in vivo metabolite patterns of 2,5-difluoroaminobenzene and of its nitrobenzene analogue, 2,5-difluoronitrobenzene, were determined using 19F NMR analysis of urine samples. Results obtained demonstrate significant differences between the biotransformation patterns of these two analogues. For the aminobenzene, cytochrome P450

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service