Skip to Content
Merck
All Photos(1)

Key Documents

302465

Sigma-Aldrich

2-Bromo-4-fluorophenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(F)OH
CAS Number:
Molecular Weight:
191.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

solid

bp

89 °C/1 mmHg (lit.)

mp

43-45 °C (lit.)

SMILES string

Oc1ccc(F)cc1Br

InChI

1S/C6H4BrFO/c7-5-3-4(8)1-2-6(5)9/h1-3,9H

InChI key

MEYRABVEYCFHHB-UHFFFAOYSA-N

Application

2-Bromo-4-fluorophenol was used in preparation of 4,4-difluoro-2-bromo-cyclohexadienone[1] and 6-(aminomethylphenoxy)benzoxaborole analogs.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsutomu Akama et al.
Bioorganic & medicinal chemistry letters, 23(6), 1680-1683 (2013-02-16)
A series of novel 6-(aminomethylphenoxy)benzoxaborole analogs was synthesized for the investigation of the structure-activity relationship of the inhibition of TNF-alpha, IL-1beta, and IL-6, from lipopolysaccharide stimulated peripheral blood mononuclear cells. Compounds 9d and 9e showed potent activity against all three
Metal-free oxidative fluorination of phenols with [18F]fluoride.
Zhanghua Gao et al.
Angewandte Chemie (International ed. in English), 51(27), 6733-6737 (2012-05-29)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service