Skip to Content
Merck
All Photos(1)

Key Documents

B59802

Sigma-Aldrich

4-Bromobutyronitrile

97%

Synonym(s):

1-Bromo-3-cyanopropane, 3-Bromopropyl cyanide, 3-Cyanopropyl bromide, 4-Bromobutyronitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€36.30
25 G
€109.00

€36.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€36.30
25 G
€109.00

About This Item

Linear Formula:
Br(CH2)3CN
CAS Number:
Molecular Weight:
148.00
Beilstein:
956577
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€36.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.478 (lit.)

bp

205 °C (lit.)

density

1.489 g/mL at 25 °C (lit.)

SMILES string

BrCCCC#N

InChI

1S/C4H6BrN/c5-3-1-2-4-6/h1-3H2

InChI key

CQPGDDAKTTWVDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojie Zhao et al.
Journal of chromatography. A, 1587, 197-208 (2018-12-26)
Ionic liquid bonded polysiloxanes (PILs) are a class of polysiloxanes whose side chains contain ionic liquid (IL) moieties. Considering their excellent selectivity and thermo-stability, PILs have great potentials in the development of polar stationary phases for gas chromatography. In this
Franziska Kupke et al.
Scientific reports, 6, 37631-37631 (2016-11-25)
Isothiocyanates are the most intensively studied breakdown products of glucosinolates from Brassica plants and well recognized for their pleiotropic effects against cancer but also for their genotoxic potential. However, knowledge about the bioactivity of glucosinolate-borne nitriles in foods is very

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service