Skip to Content
Merck
All Photos(1)

Key Documents

380350

Sigma-Aldrich

2-Bromo-5-fluorophenol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€63.10

€63.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€63.10

About This Item

Linear Formula:
BrC6H3(F)OH
CAS Number:
Molecular Weight:
191.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€63.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.553 (lit.)

density

1.717 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

Oc1cc(F)ccc1Br

InChI

1S/C6H4BrFO/c7-5-2-1-4(8)3-6(5)9/h1-3,9H

InChI key

HUVAOAVBKOVPBZ-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
344052369071528196
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

density

1.717 g/mL at 25 °C (lit.)

density

-

density

1.829 g/mL at 25 °C (lit.)

density

-

refractive index

n20/D 1.553 (lit.)

refractive index

-

refractive index

n20/D 1.531 (lit.)

refractive index

-

form

liquid

form

-

form

liquid

form

-

functional group

bromo

functional group

bromo, fluoro, nitrile

functional group

bromo, fluoro

functional group

carboxylic acid, fluoro

General description

2-Bromo-5-fluorophenol an aryl fluorinated building block.

Application

2-Bromo-5-fluorophenol may be used for the preparation of reaction intermediates required for the synthesis of polycyclic 5-HT3 and 5-HT4 antagonists.[1] It may be used for the the nucleophilic substitution of the 2,2′,4,4′-tetrabromodiphenyl iodonium chloride (iodyl salt).[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereospecific synthesis of mexiletine and related compounds: Mitsunobu versus Williamson reaction.
Carocci A, et al.
Tetrahedron Asymmetry, 11(17), 3619-3634 (2000)
Johannes Klösener et al.
Chemical research in toxicology, 22(1), 179-186 (2008-12-30)
Polybrominated diphenyl ethers (PBDEs) are flame retardants applied as coatings to many consumer products, including household items. PBDEs are released and produce airborne vapors and dusts. Inhalation of particle-phase and/or gas-phase PBDEs is therefore a major route of exposure. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service