All Photos(1)
About This Item
Empirical Formula (Hill Notation):
CH4
CAS Number:
Molecular Weight:
16.04
Beilstein:
1718732
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
0.55 (vs air)
Quality Level
Assay
≥99.0%
form
gas
autoignition temp.
998 °F
expl. lim.
15 %
bp
−161 °C (lit.)
mp
−183 °C (lit.)
density
0.716 g/mL at 25 °C (lit.)
SMILES string
[H]C([H])([H])[H]
InChI
1S/CH4/h1H4
InChI key
VNWKTOKETHGBQD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
Methane is used as a carbon source for the chemical vapor deposition (CVD) of carbide and diamond thin films.
Recommended products
Brass control valve Z146943 or either of brass regulators Z146706 and Z146714 is recommended.
recommended
Product No.
Description
Pricing
regulator
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Gas 1A - Press. Gas Compr. Gas
Storage Class Code
2A - Gases
WGK
nwg
Flash Point(F)
-306.4 °F - closed cup
Flash Point(C)
-188 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Judlyn Fernandes et al.
The Journal of nutrition, 143(8), 1269-1275 (2013-06-07)
Recent attention has focused on the significance of colonic Archaea in human health and energy metabolism. The main objectives of this study were to determine the associations among the number of fecal Archaea, body mass index (BMI), fecal short chain
Jeanette Ringvoll et al.
The EMBO journal, 25(10), 2189-2198 (2006-04-28)
Two human homologs of the Escherichia coli AlkB protein, denoted hABH2 and hABH3, were recently shown to directly reverse 1-methyladenine (1meA) and 3-methylcytosine (3meC) damages in DNA. We demonstrate that mice lacking functional mABH2 or mABH3 genes, or both, are
A N Hristov et al.
Journal of animal science, 91(11), 5045-5069 (2013-09-21)
The goal of this review was to analyze published data related to mitigation of enteric methane (CH4) emissions from ruminant animals to document the most effective and sustainable strategies. Increasing forage digestibility and digestible forage intake was one of the
A N Hristov et al.
Journal of animal science, 91(11), 5095-5113 (2013-09-21)
The goal of this review was to analyze published data on animal management practices that mitigate enteric methane (CH4) and nitrous oxide (N2O) emissions from animal operations. Increasing animal productivity can be a very effective strategy for reducing greenhouse gas
P J Gerber et al.
Animal : an international journal of animal bioscience, 7 Suppl 2, 220-234 (2013-06-14)
Although livestock production accounts for a sizeable share of global greenhouse gas emissions, numerous technical options have been identified to mitigate these emissions. In this review, a subset of these options, which have proven to be effective, are discussed. These
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service