Skip to Content
Merck
All Photos(2)

Key Documents

769126

Sigma-Aldrich

Methane

Messer® CANGas, 99.995%

Synonym(s):

Methane gas

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH4
CAS Number:
Molecular Weight:
16.04
Beilstein:
1718732
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
form:
gas
Assay:
99.995%
Pricing and availability is not currently available.

vapor density

0.55 (vs air)

Quality Level

description

Filling mass: 8g
Filling volume: 12L

Assay

99.995%

form

gas

autoignition temp.

998 °F

expl. lim.

15 %

bp

−161 °C (lit.)

mp

−183 °C (lit.)

density

0.716 g/mL at 25 °C (lit.)

SMILES string

[H]C([H])([H])[H]

InChI

1S/CH4/h1H4

InChI key

VNWKTOKETHGBQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methane is a colourless, odourless, and flammable gas that is lighter than air. Methane is used as a fuel for power generation in gas turbines and steam boilers; household cooking and heating, and for vehicles (compressed natural gas, CNG). It is also a feedstock of choice for production of hydrogen, carbon monoxide, methanol, acetic acid, and myriad organic compounds. Gas is easy to handle in a small, light pressure can, while the disposable container allows for aluminium recycling when finished.

Features and Benefits

Methane is used as a carbon source for the chemical vapor deposition (CVD) of carbide and diamond thin films.

Recommended products

Z742374: CANgas pressure regulator
Z742379: CANgas plastic hose spout
Z742378: CANgas outer adaptor
Z742377: CANgas inner adaptor
Z742375: CANgas dosing valve

Legal Information

Messer is a registered trademark of Messer Group GmbH

accessory

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-306.4 °F - closed cup

Flash Point(C)

-188 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Neil Robinson et al.
Physical chemistry chemical physics : PCCP, 22(24), 13689-13697 (2020-06-12)
Understanding the uptake and storage of gases by microporous materials is important for our future energy security. As such, we demonstrate here the application of two-dimensional NMR relaxation experiments for probing the admission and corresponding exchange dynamics of methane within

Questions

1–2 of 2 Questions  
  1. I want to buy 1L CH4 (SKU 769126) and 1L N2O (SKU 768960) to do experiment, so I also need 'mass flow controller' and 'reduction valves' that suitable to these gas cylinders. Do you have suitable 'mass flow controller' and 'reduction valves' for selling?

    1 answer
    1. Product Nos. 769126 (Methane) and 768960 (Nitrous oxide) are manufactured with proprietary Messer® CANGas canisters. The included shut-off valves in these canisters use a 7/16” 28 UNEF threaded connection.

      A compatible pressure-reducing regulator manufactured by Messer® is available under Product No. Z742374. This pressure regulator has an outlet with an NPT 1/8" female thread, a built-in gauge, and a built-in downstream dosing (metering) valve to control the flow rate. This regulator is calibrated for the 12 bar pressure of the CANGas canister, and the reduced pressure is 0.6 bar (fixed factory setting). A plastic hose barbed spout adapter is available under Product No. Z742379 for alternative outlet connections.

      A mass flow controller unfortunately is not available in our portfolio, but any mass flow controller functional at a pressure of 0.6 bar and capable of making connections with 1/8" NPT female or barbed spout outlets would be compatible.

      Helpful?

  2. Hi, I bought the 1L methane gas to connect to CI port of our new GC MS, but I do struggle to find the right regulator which can control the pressure/flow of the gas from the Can and has the outlet of 1/8''. Could you please advice

    1 answer
    1. The following regulator and accessories are recommended for regulation and flow control of this gas cylinder.

      Z742374: CANgas pressure regulator
      Z742379: CANgas plastic hose spout
      Z742378: CANgas outer adaptor
      Z742377: CANgas inner adaptor
      Z742375: CANgas dosing valve

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service