Skip to Content
Merck
All Photos(2)

Documents

289108

Sigma-Aldrich

Chloro(dimethyl)octadecylsilane

95%

Synonym(s):

Dimethyloctadecylchlorosilane, Octadecyldimethylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17Si(CH3)2Cl
CAS Number:
Molecular Weight:
347.09
Beilstein:
2246012
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

bp

145-155 °C/0.005 mmHg (lit.)

mp

28-30 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC[Si](C)(C)Cl

InChI

1S/C20H43ClSi/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-22(2,3)21/h4-20H2,1-3H3

InChI key

GZGREZWGCWVAEE-UHFFFAOYSA-N

Application

Silylating reagent

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

237.2 °F - Pensky-Martens closed cup

Flash Point(C)

114 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen Mejía-Carmona et al.
Journal of chromatography. A, 1621, 461089-461089 (2020-05-05)
Given the increasing need for analyzing natural or contaminating compounds in complex food matrices in a simple and automated way, coupling miniaturized sample preparation techniques with chromatographic systems have become a growing field of research. In this regard, given the
Faiz Ali et al.
Journal of separation science, 37(23), 3426-3434 (2014-09-23)
Ground porous silica monolith particles with an average particle size of 2.34 μm and large pores (363 Å) exhibiting excellent chromatographic performance have been synthesized on a relatively large scale by a sophisticated sol-gel procedure. The particle size distribution was
Victor K Langsi et al.
Journal of chromatography. A, 1402, 17-26 (2015-06-02)
Nanometer control over the porous shell thickness of sub-2-μm-shell particles is investigated. Three seeded growth mesoporous thin shell particles for HPLC were prepared, with 0.05μm (or 50nm) porous shell layers: particle sizes 1.5μm (solid core diameters 1.4μm), 1.7μm (solid core
Adelina Smirnova et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 31(11), 1201-1204 (2015-11-13)
Micro- and nanofluidics has attracted much attention, particularly concerning single-cell analysis when small amounts of liquids are examined. In present work we successfully fabricated extended-nano channels that were more narrow and shorter (2 mm) as well as wider and longer
Elena Ortiz-Villanueva et al.
Analytica chimica acta, 846, 51-59 (2014-09-16)
In this study, C18-silica monoliths were synthesized as a porous layer in open tubular capillary columns, to be cut later into microcartridges for the analysis of neuropeptides by on-line solid-phase extraction capillary electrophoresis with UV and MS detection (SPE-CE-UV and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service