Merck
  • Preparation and evaluation of open tubular C18-silica monolithic microcartridges for preconcentration of peptides by on-line solid phase extraction capillary electrophoresis.
Preparation and evaluation of open tubular C18-silica monolithic microcartridges for preconcentration of peptides by on-line solid phase extraction capillary electrophoresis.
Analytica chimica acta (2014-09-16)
Elena Ortiz-Villanueva, Fernando Benavente, Estela Giménez, Fatma Yilmaz, Victoria Sanz-Nebot
ABSTRACT
In this study, C18-silica monoliths were synthesized as a porous layer in open tubular capillary columns, to be cut later into microcartridges for the analysis of neuropeptides by on-line solid-phase extraction capillary electrophoresis with UV and MS detection (SPE-CE-UV and SPE-CE-MS). First, several types of C18-silica monolithic (MtC18) microcartridges were used to analyse standard solutions of five neuropeptides (i.e. dynorphin A (1-7), substance P (7-11), endomorphin 1, methionine enkephalin and [Ala]-methionine enkephalin). The MtC18 sorbents were especially selective against endomorphin 1 and substance P (7-11)). The best results in terms of sensitivity and inter-microcartridge reproducibility were achieved with the microcartridges obtained from a 10-cm open tubular capillary column with a thin monolithic coating with large through-pores (1-5μm). Run-to-run repeatability, microcartridge durability, linearity ranges and LODs were studied by MtC18-SPE-CE-MS. As expected due to their greater selectivity, the best LOD enhancement was obtained for End1 and SP (7-11) (50 times with regard to CE-MS). Finally, the suitability of the methodology for analysing biological fluids was tested with plasma samples spiked with End1 and SP (7-11). Results obtained were promising because both neuropeptides could be detected at 0.05μgmL(-1), which was almost the same concentration level as for the standard solutions (0.01μgmL(-1)).
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetic acid, BioUltra, ≥99.5% (GC)
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Methanol, analytical standard
Supelco
Acetone, analytical standard
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, ≥98.0% (T), pellets
Sigma-Aldrich
Chloro(dimethyl)octadecylsilane, 95%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Ammonia solution, 4 M in methanol