Skip to Content
Merck
All Photos(1)

Key Documents

246859

Sigma-Aldrich

Chloro(dimethyl)octylsilane

97%

Synonym(s):

Dimethyloctylchlorosilane, Octyldimethylchlorosilane

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€135.00

€135.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€135.00

About This Item

Linear Formula:
CH3(CH2)7Si(CH3)2Cl
CAS Number:
Molecular Weight:
206.83
Beilstein:
1738376
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

€135.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.435 (lit.)

bp

222-225 °C (lit.)

density

0.873 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC[Si](C)(C)Cl

InChI

1S/C10H23ClSi/c1-4-5-6-7-8-9-10-12(2,3)11/h4-10H2,1-3H3

InChI key

DBKNGKYVNBJWHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloro(dimethyl)octylsilane is an organosilicon compound that is primarily used as a reagent in organic synthesis for the functionalization of the surfaces of various materials, such as silica, glass, and polymers. It is also often employed as a precursor in the synthesis of various organosilicon compounds.

Application

Chloro(dimethyl)octylsilane can be used as a reagent for the silylation of various materials, which includes silica and graphite.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorena González et al.
Journal of chromatography. A, 1622, 461124-461124 (2020-05-08)
In this work a new method for the determination of 5-hydroxymethylfurfural (HMF) in cereal and insect bars has been developed and validated. The method consisted of a solid-liquid extraction (SLE) followed by a solid phase extraction (SPE), employing functionalized mesostructured
Polina Naidjonoka et al.
Soft matter, 16(30), 7063-7076 (2020-08-07)
The substantial part of the water-soluble hemicellulose fraction, obtained when processing cellulose to produce paper and other products, has so far been discarded. The aim of this work is to reveal the interfacial properties of softwood hemicellulose (galactoglucomannan, GGM) in
Natalia Casado et al.
Talanta, 165, 223-230 (2017-02-06)
A SBA-15 type mesoporous silica was synthesized and bi-functionalized with octadecylsilane (C18) or octylsilane (C8), and sulfonic acid (SO

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service