Skip to Content
Merck
All Photos(1)

Key Documents

287482

Sigma-Aldrich

4-Octylbenzenesulfonic acid sodium salt

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€184.00

€184.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€184.00

About This Item

Linear Formula:
CH3(CH2)7C6H4SO3Na
CAS Number:
Molecular Weight:
292.37
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€184.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

powder

functional group

sulfonic acid

SMILES string

[Na+].CCCCCCCCc1ccc(cc1)S([O-])(=O)=O

InChI

1S/C14H22O3S.Na/c1-2-3-4-5-6-7-8-13-9-11-14(12-10-13)18(15,16)17;/h9-12H,2-8H2,1H3,(H,15,16,17);/q;+1/p-1

InChI key

ASJUTVUXOMPOQH-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dale R Van Stempvoort et al.
Chemosphere, 251, 126386-126386 (2020-03-11)
This study presents the first reported detections and concentrations of alkyl phenoxy-benzenesulfonate surfactants (APBS) in municipal wastewater. A semi quantitative direct injection LC/MS/MS method was developed. Samples of raw influent and final effluent were obtained from fourteen municipal wastewater treatment
Stephanie R Vaughan et al.
Sensors (Basel, Switzerland), 20(3) (2020-01-26)
Herein, we examine two sensing schemes for detection and discrimination of chlorinated volatile organic compounds (VOCs). In this work, phosphonium ionic liquids (ILs) were synthesized and vapor sensing properties examined and compared to phosphonium IL-polymer composites. Pure IL sensors were
Patrik Knöös et al.
International journal of pharmaceutics, 468(1-2), 121-132 (2014-04-15)
The release of lactose (hydrophilic) from polymer tablets made with hydrophobically modified poly(acrylic acid) (HMPAA) have been studied and compared to the release of ibuprofen, a hydrophobic active substance. Lactose is one of the most used excipients for tablets, but

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service