Skip to Content
Merck
All Photos(2)

Key Documents

522988

Sigma-Aldrich

Dinonylnaphthalenedisulfonic acid solution

55 wt. % in isobutanol

Synonym(s):

DNNDSA

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€233.00

€233.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€233.00

About This Item

Linear Formula:
(C9H19)2C10H4(SO3H)2
CAS Number:
Molecular Weight:
540.78
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.23

€233.00


Check Cart for Availability

Request a Bulk Order

Quality Level

concentration

55 wt. % in isobutanol

refractive index

n20/D 1.476

bp

101 °C

solubility

H2O: insoluble
alcohols, glycol ethers, glycols, esters, ketones, aromatic and aliphatic hydrocarbons: soluble

density

0.98 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCCCCCCCc1ccc2c(c(CCCCCCCCC)ccc2c1S(O)(=O)=O)S(O)(=O)=O

InChI

1S/C28H44O6S2/c1-3-5-7-9-11-13-15-17-23-19-21-26-25(27(23)35(29,30)31)22-20-24(28(26)36(32,33)34)18-16-14-12-10-8-6-4-2/h19-22H,3-18H2,1-2H3,(H,29,30,31)(H,32,33,34)

InChI key

KVOYCYJHAKSFSR-UHFFFAOYSA-N

General description

Dinonylnaphthalenedisulfonic acid solution (DNNDSA) is an aromatic sulphonic acid, hydrophobic in nature.2

Application

Dopant for conducting polymers & hydrophobic acid catalyst for amino cross-linked coatings

Packaging

Packaged in glass bottles

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

115.0 °F - closed cup

Flash Point(C)

46.11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of polyaniline nanoparticles in SDS micellar solutions
Kim B, et al.
Synt. Metals, 122(2), 297-304 (2001)
Tarick J El-Baba et al.
Rapid communications in mass spectrometry : RCM, 28(11), 1175-1184 (2014-04-25)
New inlet and vacuum ionization methods provide advantages of specificity, simplicity and speed for the analysis of synthetic polymers and polymer additives directly from surfaces such as fibers using mass spectrometry (MS) on different commercial mass spectrometers (Waters SYNAPT G2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service