Merck
  • Quantifying the release of lactose from polymer matrix tablets with an amperometric biosensor utilizing cellobiose dehydrogenase.
Quantifying the release of lactose from polymer matrix tablets with an amperometric biosensor utilizing cellobiose dehydrogenase.
International journal of pharmaceutics (2014-04-15)
Patrik Knöös, Christopher Schulz, Lennart Piculell, Roland Ludwig, Lo Gorton, Marie Wahlgren
ABSTRACT
The release of lactose (hydrophilic) from polymer tablets made with hydrophobically modified poly(acrylic acid) (HMPAA) have been studied and compared to the release of ibuprofen, a hydrophobic active substance. Lactose is one of the most used excipients for tablets, but lactose release has not been widely studied. One reason could be a lack of good analytical tools. A novel biosensor with cellobiose dehydrogenase (CDH) was used to detect the lactose release, which has a polydiallyldimethylammonium chloride (PDADMAC) layer that increases the response. A sample treatment using polyethylenimine (PEI) was developed to eliminate possible denaturants. The developed methodology provided a good approach to detect and quantify the released lactose. The release was studied with or without the presence of a model amphiphilic substance, sodium dodecyl sulphate (SDS), in the release medium. Ibuprofen showed very different release rates in the different media, which was attributed to hydrophobic interactions between the drug, the HMPAA and the SDS in the release medium. The release of hydrophilic lactose, which did not associate to any of the other components, was rapid and showed only minor differences. The new methodology provides a useful tool to further evaluate tablet formulations by a relatively simple set of experiments.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Supelco
Ibuprofen sodium salt, ≥98% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sigma-Aldrich
4-Octylbenzenesulfonic acid sodium salt, 97%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard