Skip to Content
Merck
All Photos(1)

Key Documents

283606

Sigma-Aldrich

Palladium(II) chloride solution

5 wt. % in 10 wt. % HCl

Synonym(s):

Dichloropalladium, Palladium dichloride, Palladous chloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€434.00

€434.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
€434.00

About This Item

Linear Formula:
PdCl2
CAS Number:
Molecular Weight:
177.33
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€434.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

concentration

5 wt. % in 10 wt. % HCl

density

1.06 g/mL at 25 °C

SMILES string

Cl[Pd]Cl

InChI

1S/2ClH.Pd/h2*1H;/q;;+2/p-2

InChI key

PIBWKRNGBLPSSY-UHFFFAOYSA-L

General description

Palladium(II) chloride is a representative salt of Pd(II) that is used as a precursor to prepare palladium catalysts for various reactions, such as Heck coupling, Suzuki coupling, Sonagashira coupling, and Buchwald-Hartwig coupling. It is also used as an oxidizing agent.

Application

Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathis, M. et al.
Chemistry of Materials, 10, 3568-3568 (1998)
Yves Canac et al.
Inorganic chemistry, 50(21), 10810-10819 (2011-10-11)
The influence of the formal electrostatic interaction on the cis/trans coordination mode at a PdCl(2) center is investigated in a family of isostructural flexible diphosphine ligands Ph(2)P-X-C(6)H(4)-Y-PPh(2), where X and Y stand for neutral or cationic N,C-imidazolylene linkers. While the
Buyi Li et al.
Advanced materials (Deerfield Beach, Fla.), 24(25), 3390-3395 (2012-06-08)
Highly dispersed palladium chloride catalysts locked in triphenylphosphine-functionalized knitting aryl network polymers (KAPs) are developed and exhibit excellent activity under mild conditions in the Suzuki-Miyaura cross-coupling reactions of aryl chlorides in aqueous media. This work highlights that the microporous polymers
Maozhong Miao et al.
Organic letters, 14(11), 2718-2721 (2012-05-16)
A novel PdCl(2)-catalyzed oxidative cycloisomerization of 3-cyclopropylideneprop-2-en-1-ones, providing a facile synthesis of highly strained functionalized 2-alkylidenecyclobutanones via furan-fused cyclobutene intermediates, is reported. An interesting route to 2(3H)-furanones with a spiro-cyclopropane unit from the obtained 2-alkylidenecyclobutanones via a ring-contraction rearrangement reaction
Juraj Velcicky et al.
Journal of the American Chemical Society, 133(18), 6948-6951 (2011-04-21)
A one-pot protocol for the cyanomethylation of aryl halides through a palladium-catalyzed reaction with isoxazole-4-boronic acid pinacol ester was developed. Mechanistically, the reaction proceeds through (1) Suzuki coupling, (2) base-induced fragmentation, and (3) deformylation as shown by characterization of all

Questions

  1. Does the 5 wt. % in 10 wt. % HCl mean PdCl2 weight percent or just the Pd weight percent? i.e if the solution weight is 100g, is the mass of Pd 5g or mass of PdCl2 5g?

    1 answer
    1. 5 wt% is the weight of PdCl2. If the solution weight is 100g, the mass of PdCl2 is 5g.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service