Skip to Content
Merck
All Photos(1)

Key Documents

76050

Sigma-Aldrich

Palladium(II) chloride

anhydrous, 60% Pd basis

Synonym(s):

Dichloropalladium, Palladium dichloride, Palladous chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€131.00
5 G
€622.00
25 G
€3,040.00

€131.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€131.00
5 G
€622.00
25 G
€3,040.00

About This Item

Empirical Formula (Hill Notation):
Cl2Pd
CAS Number:
Molecular Weight:
177.33
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€131.00


Check Cart for Availability

Request a Bulk Order

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

concentration

60% Pd

mp

678-680 °C (lit.)

density

4 g/mL at 25 °C (lit.)

SMILES string

Cl[Pd]Cl

InChI

1S/2ClH.Pd/h2*1H;/q;;+2/p-2

InChI key

PIBWKRNGBLPSSY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Application Guide for Palladium Catalyzed Cross-Coupling Reactions

Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.
Palladium(II) chloride can be used to catalyze:
  • Cross-coupling reactions between terminal alkynes and aryl iodides or bromides under modified Sonogashira-Cassar-Heck conditions.[1]
  • Carbonylation of organic tellurides with carbon monoxide to form corresponding methyl carboxylates.[2]
  • Isomerization of allylic ester in acetic acid.[3]
  • Carbonylation of amines to form isocyanates.[4]

Other Notes

Review[5][6]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium (II)-catalyzed exchange and isomerization reactions. III. Allylic esters isomerization in acetic acid catalyzed by palladium (II) chloride.
Henry P M.
Journal of the American Chemical Society, 94(15), 5200-5206 (1972)
Carbonylation of Amines in the Presence of Palladium (II) Chloride. A New Route to Isocyanates.
Stern E. W and Spector M. L.
The Journal of Organic Chemistry, 31(2), 596-597 (1966)
Palladium (II) Chloride and a (Dipyridin?2?ylmethyl) amine?Derived Palladium (II) Chloride Complex as Highly Efficient Catalysts for the Synthesis of Alkynes in Water or in NMP and of Diynes in the Absence of Reoxidant.
Gil?Molto J and Najera C.
European Journal of Organic Chemistry, 2005(19), 4073-4081 (2005)
R.F. Heck
Accounts of Chemical Research, 12, 146-146 (1979)
Palladium (II) chloride catalyzed carbonylation of organic tellurides with carbon monoxide.
Ohe K, et al.
The Journal of Organic Chemistry, 52(22), 4859-4863 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service